Anti-DPP7 Mouse Polyclonal Antibody
Catalog # ABNOH00029952-B01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Dipeptidylpeptidase 7
- Antigen symbol:DPP7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:29952
- Antigen synonyms:QPP|DPP2|DPPII
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: DPP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human