Anti-WAPAL Mouse Polyclonal Antibody
Catalog # ABNOH00023063-B01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Wings Apart-Like
- Antigen symbol:WAPAL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:23063
- Antigen synonyms:FOE|Wings apart-like protein homolog|KIAA0261|WAPL|Friend of EBNA2 protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: WAPAL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human