Anti-P3H4 Mouse Polyclonal Antibody
ABNOH00010609-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Prolyl 3-Hydroxylase 4
- Antigen symbol:P3H4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10609
- Antigen synonyms:SC65|LEPREL4|NO55|NOL55
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: P3H4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human