Order Entry
Singapore
ContactUsLinkComponent
Anti-CHST4 Mouse Monoclonal Antibody [clone: 4D7]
Anti-CHST4 Mouse Monoclonal Antibody [clone: 4D7]
Catalog # ABNOH00010164-M10
Supplier:  Abnova
undefined
Anti-CHST4 Mouse Monoclonal Antibody [clone: 4D7]
Catalog # ABNOH00010164-M10
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Carbohydrate sulfotransferase 4
  • Antigen symbol:
    CHST4
  • Clonality:
    Monoclonal
  • Clone:
    4D7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    10164
  • Antigen synonyms:
    Gn6st-2|GST-3|LSST|HEC-GlcNAc6ST
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: CHST4
Clonality: Monoclonal
Clone: 4D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human