Anti-AIMP1 Mouse Monoclonal Antibody [clone: M1]
ABNOH00009255-M02
: Abnova
- Antibody type:Primary
- Antigen name:Aminoacyl TRNA Synthetase Complex-Interacting Multifunctional Protein 1
- Antigen symbol:AIMP1
- Clonality:Monoclonal
- Clone:M1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:9255
- Antigen synonyms:HLD3|p43|EMAPII|SCYE1|EMAP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: AIMP1
Clonality: Monoclonal
Clone: M1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human