Anti-AAAS Mouse Polyclonal Antibody
Catalog # ABNOH00008086-B01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Adracalin
- Antigen symbol:AAAS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8086
- Antigen synonyms:AAASb|DKFZp586G1624|ADRACALA|Aladin|Allgrove triple A|AAAS_HUMAN.|GL003|AAAS|AAA|Achalasia adrenocortical insufficiency alacrimia
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: AAAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human