Order Entry
Singapore
ContactUsLinkComponent
Anti-RNF103 Mouse Monoclonal Antibody [clone: 1C1]
Anti-RNF103 Mouse Monoclonal Antibody [clone: 1C1]
Catalog # ABNOH00007844-M11
Supplier:  Abnova
undefined
Anti-RNF103 Mouse Monoclonal Antibody [clone: 1C1]
Catalog # ABNOH00007844-M11
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ring Finger Protein 103
  • Antigen symbol:
    RNF103
  • Clonality:
    Monoclonal
  • Clone:
    1C1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    7844
  • Antigen synonyms:
    AW212918|Zfp103|AW146237|kf-1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: RNF103
Clonality: Monoclonal
Clone: 1C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human