Order Entry
Singapore
ContactUsLinkComponent
Anti-TPM2 Mouse Monoclonal Antibody [clone: 3C8]
Anti-TPM2 Mouse Monoclonal Antibody [clone: 3C8]
  ABNOH00007169-M07
 :  Abnova
undefined
Anti-TPM2 Mouse Monoclonal Antibody [clone: 3C8]
  ABNOH00007169-M07
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Tropomyosin 2
  • Antigen symbol:
    TPM2
  • Clonality:
    Monoclonal
  • Clone:
    3C8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    7169
  • Antigen synonyms:
    DA1|HEL-S-273|DA2B|TMSB|AMCD1|NEM4
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: TPM2
Clonality: Monoclonal
Clone: 3C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human