Anti-SGCA Mouse Monoclonal Antibody [clone: 3C4]
ABNOH00006442-M01
: Abnova
- Antibody type:Primary
- Antigen name:Sarcoglycan alpha (50kDa dystrophin-associated glycoprotein)
- Antigen symbol:SGCA
- Clonality:Monoclonal
- Clone:3C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6442
- Antigen synonyms:ADL|DAG2|DMDA2|50-DAG|adhalin|SCARMD1|A2|LGMD2D
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SGCA
Clonality: Monoclonal
Clone: 3C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human