Anti-S100 A5 Mouse Monoclonal Antibody [clone: 5E1]
Catalog # ABNOH00006276-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:S100 Calcium binding Protein A5
- Antigen symbol:S100 A5
- Clonality:Monoclonal
- Clone:5E1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6276
- Antigen synonyms:S100D
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: S100 A5
Clonality: Monoclonal
Clone: 5E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human