Anti-RPS9 Mouse Polyclonal Antibody
ABNOH00006203-A01
: Abnova
- Antibody type:Primary
- Antigen name:Ribosomal Protein 9
- Antigen symbol:RPS9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6203
- Antigen synonyms:40S ribosomal protein S9|RPS9
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: RPS9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat