Order Entry
Singapore
ContactUsLinkComponent
 
Anti-PPOX Mouse Monoclonal Antibody [clone: 1E1]
  ABNOH00005498-M06
 :  Abnova
undefined
Anti-PPOX Mouse Monoclonal Antibody [clone: 1E1]
  ABNOH00005498-M06
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Protoporphyrinogen Oxidase
  • Antigen symbol:
    PPOX
  • Clonality:
    Monoclonal
  • Clone:
    1E1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    5498
  • Antigen synonyms:
    V290M|VP|PPO
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: PPOX
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human