Anti-MBNL1 Mouse Monoclonal Antibody [clone: 1D11]
ABNOH00004154-M01J
: Abnova
- Antibody type:Primary
- Antigen name:Muscleblind-like splicing regulator 1
- Antigen symbol:MBNL1
- Clonality:Monoclonal
- Clone:1D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4154
- Antigen synonyms:EXP40|muscleblind-like (Drosophila)|KIAA0428|MBNL1|muscleblind-like 1|EXP42|EXP|MBNL|EXP35|DKFZp686P06174
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: MBNL1
Clonality: Monoclonal
Clone: 1D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human