Order Entry
Singapore
ContactUsLinkComponent
Anti-BRF1 Mouse Monoclonal Antibody [clone: 2E9]
Anti-BRF1 Mouse Monoclonal Antibody [clone: 2E9]
  ABNOH00002972-M01
 :  Abnova
undefined
Anti-BRF1 Mouse Monoclonal Antibody [clone: 2E9]
  ABNOH00002972-M01
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit
  • Antigen symbol:
    BRF1
  • Clonality:
    Monoclonal
  • Clone:
    2E9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    2972
  • Antigen synonyms:
    HEL-S-76p|TFIIIB90|TAF3B2|TAFIII90|GTF3B|hBRF|RF|TF3B90|RNA polymerase III transcription initiation factor 90 kDa subunit|TAF3C|BRF-1|CFDS
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µg

 

 

Type: Primary
Antigen: BRF1
Clonality: Monoclonal
Clone: 2E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human