Anti-KAT2A Mouse Monoclonal Antibody [clone: 4D3]
ABNOH00002648-M01
: Abnova
- Antibody type:Primary
- Antigen name:K(lysine) acetyltransferase 2A
- Antigen symbol:KAT2A
- Clonality:Monoclonal
- Clone:4D3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2648
- Antigen synonyms:hGCN5|PCAF-b|GCN5L2|GCN5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: KAT2A
Clonality: Monoclonal
Clone: 4D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human