Order Entry
Singapore
ContactUsLinkComponent
 
Anti-EFNA1 Rabbit Polyclonal Antibody
  ABNOH00001942-D01P
 :  Abnova
undefined
Anti-EFNA1 Rabbit Polyclonal Antibody
  ABNOH00001942-D01P
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Ephrin A1
  • Antigen symbol:
    EFNA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    1942
  • Antigen synonyms:
    tumor necrosis factor|ligand of eph-related kinase 1|TNFAIP4|eph-related receptor tyrosine kinase ligand 1|ECKLG|EFL1|alpha-induced protein 4|immediate early response protein B61|B61|EPLG1|ephrin A1|ephrin-A1|EFNA1|LERK1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: EFNA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human