Anti-EEF1D Mouse Monoclonal Antibody [clone: 4B12]
Catalog # ABNOH00001936-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Elongation factor 1-delta
- Antigen symbol:EEF1D
- Clonality:Monoclonal
- Clone:4B12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1936
- Antigen synonyms:FP1047|EF-1D|EF1D
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: EEF1D
Clonality: Monoclonal
Clone: 4B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human