Anti-DGKA Mouse Monoclonal Antibody [clone: 2B7]
Catalog # ABNOH00001606-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Diacylglycerol kinase, alpha 80kDa
- Antigen symbol:DGKA
- Clonality:Monoclonal
- Clone:2B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1606
- Antigen synonyms:DAGK|80 kDa diacylglycerol kinase|Diacylglycerol kinase alpha|DAG kinase alpha|DAGK1|DGK-alpha|Diglyceride kinase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: DGKA
Clonality: Monoclonal
Clone: 2B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human