Respirator filter cartridges
Catalog # NORH75SC
Supplier: Honeywell Safety
Specifications
- Description:Defender® multipurpose cartridge, approved for use against organic vapour, chlorine, hydrogen chloride, sulfur dioxide, hydrogen sulfide (escape), hydrogen fluoride, chlorine dioxide, ammonia, methylamine, formaldehyde
- Environmentally Preferable:
- Pk:2
Specifications
About this item
For use with North safety products air purifying dual cartridge respirators. Adapter assembly attaches pancake filter to a gas and vapour cartridges and attaches to chemical cartridges except for the Defender® multipurpose cartridge.
- NIOSH approved when used with Nort Safety Products 7700, 7600, 5500, and 5400 air purifying respirators