Order Entry
Singapore
ContactUsLinkComponent
 
VWR® shiroGEL horizontal 96, Horizontal Electrophoresis Apparatus
  VWRI730-0340
undefined
VWR® shiroGEL horizontal 96, Horizontal Electrophoresis Apparatus
  VWRI730-0340

 

  • Description:
    shiroGEL horizontal 96 system, including comb block (12×8-sample, assembled, comb thickness 1,0 mm, 1 marker lane)
  • Environmentally Preferable:
  • Model:
    shiroGEL horizontal 96 (1 mm, 1 marker)
  • Buffer volume:
    300 ml
  • Gel size W×L:
    100×120 mm
  • Sample capacity:
    96 samples plus 1 marker lane
  • Unit W×L×H:
    220×125×90 mm
  • Pk:
    1

 

 

This 96-well format horizontal gel electrophoresis unit is compatible with 8-channel multichannel pipettes, and matches up with a standard 96-well plate configuration.

  • Average run time is 15 to 30 min per gel
  • Layout of wells allows a run length of 18 mm per sample, providing ideal separation

Combs for shiroGEL horizontal 96