Self Seal Sterilisation Pouches
Supplier: CARDINAL HEALTH
Certificates
About this item
These sterilization pouch combines a blue film and a medical grade paper to provide breathability, sterile barrier and indication of processing. To open, a convenient thumb notch is positioned at the top of the pouch, above the chevron seal. Each pouch includes an external process indicator for steam and ethylene oxide (EO) gas sterilisation.
- Pouched sealed mechanically or manually with a heat sealer
- For use in steam and gas (EO) sterilisers