3-Amino-4-bromo-5-methylisoxazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-bromo-5-methylisoxazol-3-amine
CAS Number:5819-40-9
:
Formula:C₄H₅BrN₂O
MDL Number:MFCD00052553
Molecular Weight:177.02 g/mol
Melting Pt:73…75 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: