Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(4-Chlorophenyl)methanesulphonyl chloride
(4-Chlorophenyl)methanesulphonyl chloride ≥97%
(4-Chlorophenyl)methanesulphonyl chloride ≥97%
:
Thermo Fisher Scientific
(4-Chlorophenyl)methanesulphonyl chloride ≥97%
:
Thermo Fisher Scientific
:
4-Chloro-α-toluenesulphonyl chloride
:
6966-45-6
:
:
C₇H₆Cl₂O₂S
:
MFCD01631927
:
225.10 g/mol
:
93…96 °C
:
8 II
:
GHS05
:
Ambient
Environmentally Preferable:
msds_ALFAH26017
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years