1,1,1,3,3,3-Hexafluoroisopropyl acrylate ≥98% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,1,1,3,3,3-Hexafluoropropan-2-yl acrylate
CAS Number:2160-89-6
:
Formula:C₆H₄F₆O₂
MDL Number:MFCD00040104
Boiling Pt:81…82 °C
Density:1.370
:Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
Molecular Weight:222.09 g/mol
Flash Pt:17 °C (62 °F)
Storage Temperature:Ambient
EINECS:218-479-1
UN:3272
ADR:3 II
:GHS02
:GHS07
Specifications
- Environmentally Preferable: