L(-)-Glutathione (reduced form) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced
CAS Number:70-18-8
:
Formula:C₁₀H₁₇N₃O₆S
MDL Number:MFCD00065939
Density:1.475 g/cm³ (20 °C)
Molecular Weight:307.33 g/mol
Melting Pt:∼ 193 °C (dec.)
Merck Index:13,04488
Storage Temperature:Refrigerator
EINECS:200-725-4
Specifications
- Environmentally Preferable: