4'-Fluoro-2'-methoxyacetophenone ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-(4-Fluoro-2-methoxyphenyl)ethanone
CAS Number:51788-80-8
:
Formula:C₉H₉FO₂
MDL Number:MFCD00061143
Molecular Weight:168.17 g/mol
Melting Pt:51…53 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: