Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-6,7-dimethoxyquinazoline
4-Chloro-6,7-dimethoxyquinazoline ≥98%
4-Chloro-6,7-dimethoxyquinazoline ≥98%
:
Thermo Fisher Scientific
4-Chloro-6,7-dimethoxyquinazoline ≥98%
:
Thermo Fisher Scientific
:
13790-39-1
:
:
C₁₀H₉ClN₂O₂
:
MFCD01570172
:
224.65 g/mol
:
184…187 °C
:
GHS07
:
Ambient
Environmentally Preferable:
msds_ALFAH28405
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years