Order Entry
Singapore
ContactUsLinkComponent
2-Chloro-3-fluoro-4-nitropyridine-N-oxide 95%
2-Chloro-3-fluoro-4-nitropyridine-N-oxide 95%
Catalog # ALFAL20025.03
CAS Number:  
2-Chloro-3-fluoro-4-nitropyridine-N-oxide 95%
Catalog # ALFAL20025.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :192.53 g/mol
EINECS:238-404-6
UN:2811
 :GHS06
 :GHS07
Storage Temperature:Ambient
CAS Number:101664-56-6
Formula:C₅H₂ClFN₂O₃
MDL Number:MFCD04039346
Melting Pt:176…178 °C
ADR:6.1 III