Order Entry
Singapore
ContactUsLinkComponent
Optical adhesive PCR/qPCR film for Piko® UTW plates
Optical adhesive PCR/qPCR film for Piko® UTW plates
Catalog # ADVAASF-0020/100
undefined
Optical adhesive PCR/qPCR film for Piko® UTW plates
Catalog # ADVAASF-0020/100
Specifications
  • Optical property:
    Optically clear
  • Description:
    Optical adhesive film for PCR/qPCR
  • Seq:
    01
  • Environmentally Preferable:
  • Pk:
    100
Specifications
About this item
Clear, 24-well format Piko® PCR/qPCR film seal allows high transmission of fluorescence across the range of commonly used fluorophores. When used with a Piko® thermal cycler or PikoReal® Real-Time PCR system, sample volumes as low as 5 µl can be used with minimal evaporative loss.
  • Optical clarity with minimal auto-fluorescence for Real-Time PCR applications
  • Tight sealing for sample volumes as low as 5 µl
  • Adhesive backing allows easy application without costly sealing devices
  • Cryo-compatible adhesive is effective to −80 °C