Order Entry
Portugal
ContactUsLinkComponent
 
2-Methyl-3-hexanone ≥98.0% (by GC)
  TCIAM0570-5G
 :  TCI
 :  
2-Methyl-3-hexanone ≥98.0% (by GC)
  TCIAM0570-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :114.19 g/mol
 :230-940-9
 :1224
 :GHS02
 :GHS07
 :7379-12-6
 :C₇H₁₄O
 :MFCD00009399
 :136°C
 :0.8428
 :24°C
 :3 III