2,4-Dibromophenol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:615-58-7
:
Formula:C₆H₄Br₂O
MDL Number:MFCD00002149
Boiling Pt:239 °C
Melting Pt:35…38 °C
Density:2.07 g/cm³ (20 °C)
Molecular Weight:251.91 g/mol
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:210-436-5
UN:2811
ADR:6.1 II
:GHS06
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together