Triethylammonium chloride 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Triethylamine hydrochloride
CAS Number:554-68-7
:
Formula:C₆H₁₆ClN
MDL Number:MFCD00012500
Molecular Weight:137.65 g/mol
Melting Pt:254…260 °C
EINECS:209-067-2
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:254 to 260 °C
- Loss on drying:≤1.0 %
- Solubility:(10 % in water) Clear colorless
- pH:4.5 to 6.5 (1 % w/v solution)
- Titration with HClO4:99.0 to 101.0 % (On anhydrous substance)
Specification Test Results
Frequently Bought Together