Iodine 99.5%, extra pure, sublimated
:7553-56-2
:
:I₂
:MFCD00011355
:184.3
:253.81 g/mol
:113 °C
:8 III
:13,05036
:231-442-4
:3495
:GHS07
:GHS08
:GHS09
:Ambient
- Environmentally Preferable:
- Appearance:Dark grey Metallic spheres
- Titration Iodometric:≥99.4 %
- Chloride and Bromide:≤0.005 %
- Insoluble matter:Passes test
Frequently Bought Together