Secnidazole 98%
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
Synonyms: 1-(2-Methyl-5-nitroimidazol-1-yl)-2-propanol, 1-(2-Methyl-5-nitro-1H-imidazol-1-yl)propan-2-ol
CAS Number:3366-95-8
:
Formula:C₇H₁₁N₃O₃
MDL Number:MFCD00864656
Molecular Weight:185.18 g/mol
Merck Index:13,08494
Storage Temperature:Refrigerator
EINECS:222-134-0
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:white to light yellow powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Melting point:70°C to 76°C
- Water:≤6 %
Specification Test Results
Frequently Bought Together