Nadifloxacin ≥98%
Catalog # ENZOALX380294G001
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: 9-Fluoro-8-(4-hydroxypiperidin-1-yl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic acid, 9-Fluoro-8-(4-hydroxy-1-piperidinyl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic Acid
:
:360.39 g/mol
Storage Temperature:Freezer
CAS Number:124858-35-1
Formula:C₁₉H₂₁FN₂O₄
MDL Number:MFCD00865081