4-Bromo-2,3,5,6-tetrafluorobenzoic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4707-24-8
:
Formula:C₇HBrF₄O₂
MDL Number:MFCD00075244
Molecular Weight:272.98 g/mol
Melting Pt:144…145 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together