tert-Butyl-2-(methylamino)acetate hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Sar-OtBu·HCl, tert-Butyl sarcosinate hydrochloride, Sarcosine tert-butyl ester hydrochloride, H-Sar-OtBu·HCl, tert-Butyl 2-(methylamino)acetate hydrochloride, H-Sar-OtBu.HCl
CAS Number:136088-69-2
:5616-81-9
Formula:C₇H₁₆ClNO₂
MDL Number:MFCD00080936
Molecular Weight:181.660 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together