3-Allylsalicylaldehyde
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-AllylSalicylaldehyde, 3-Allyl-2-hydroxybenzaldehyde
CAS Number:24019-66-7
:
Formula:C₁₀H₁₀O₂
MDL Number:MFCD00037364
Density:1.099 g/cm³ (25 °C)
Molecular Weight:162.19 g/mol
Melting Pt:45 °C
Flash Pt:106.7 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together