3-Amino-2-bromo-6-methoxypyridine
Catalog # APOSOR315074-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2-Bromo-6-methoxypyridin-3-amine
:
:203.0400 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:135795-46-9
Formula:C₆H₇BrN₂O
MDL Number:MFCD05662394
Melting Pt:44…45 °C