Fmoc-N-amido-dPEG4-acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:557756-85-1
:
Formula:C₂₆H₃₃NO₈
Molecular Weight:487.54 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together