Potassium iodate solution 0.05 M (0.3 N), AVS TITRINORM
Catalog # 86195.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:PE bottle, black
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Titer (20 °C):0.9980 – 1.0020
Specification Test Results
About this item
:
:214 g/mol
Storage Temperature:Ambient
CAS Number:7758-05-6
Formula:KlO₃
MDL Number:MFCD00011406
Melting Pt:560
Merck Index:13,07726