Tissue-clearing reagent, TOMEI for plants
Catalog # TCIAT3530-100ML
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mL
- Description:Tissue-Clearing reagent, TOMEI for Plants
- Environmentally Preferable:
Specifications
About this item
Specifications
Appearance: Colorless to Light yellow clear liquid
Functionality test (transparency) (display): to pass test