Chlorobis(cyclooctene)rhodium(I) dimer ≥96.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:12279-09-3
:
Formula:C₃₂H₅₆Cl₂Rh₂
MDL Number:MFCD00013287
Molecular Weight:717.51 g/mol
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (Argentmetric Titration):min. 96.0 %
Specification Test Results
Related Information
Frequently Bought Together