5-Bromo-2,4-dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:32246-20-1
:
Formula:C₉H₉BrO₄
MDL Number:MFCD05865172
Molecular Weight:261.071 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together