(S)-2-((tert-Butoxycarbonyl)amino)-3-(pyridin-2-yl)propanoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: N-Boc-3-(2-pyridyl)-L-alanine, Boc-2-Pal-OH
CAS Number:71239-85-5
:
Formula:C₁₃H₁₈N₂O₄
MDL Number:MFCD00191190
Molecular Weight:266.3 g/mol
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-15.0 to -18.0 deg (C=1, MeOH)
Specification Test Results
Frequently Bought Together