Ethyl-3-mercaptopropionate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: Ethyl 3-mercaptopropanoate, 3-Mercaptopropionic acid ethyl ester
CAS Number:5466-06-8
:
Formula:C₅H₁₀O₂S
MDL Number:MFCD00004896
Boiling Pt:75
Molecular Weight:134.2 g/mol
Flash Pt:72°C
EINECS:226-771-5
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together