N,N-Diethylformamide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Formic acid diethylamide, Formdiethylamide
CAS Number:617-84-5
:
Formula:C₅H₁₁NO
MDL Number:MFCD00003287
Boiling Pt:176
Density:0.908 g/cm³ (20 °C)
Molecular Weight:101.15 g/mol
Flash Pt:65
:M-Clarity™ quality level = MQ200
EINECS:210-533-2
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:1000 g/l (20 °C)
- Molar Mass:101.15 g/mol
- Boiling Point:178 °C
- Flash Point:65 °C
- Refractive Index:1.434 (20 °C, 589 nm)
- Density:0.91 g/cm3 (20 °C)
- Synonyms:Formic acid diethylamide
- Hill Formula:C5H11NO
- Chemical Formula:HCON(C2H5)2
Specification Test Results
Frequently Bought Together