Copper(II) sulphate pentahydrate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate
CAS Number:7758-99-8
:
Formula:CuSO₄·5H₂O
MDL Number:MFCD00149681
Density:2.284 g/cm³ (20 °C)
Molecular Weight:249.69 g/mol
ADR:9 III
Merck Index:13,02682
EINECS:231-847-6
UN:3077
REACH:01-2119520566-40
:GHS07
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 98 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 5L Plastic bucket UN Z (ZPACK13247005)
Packaging Information
spec_VWRC23165.298 spec_VWRC23165.367 spec_VWRC23165.460
Product specification
Frequently Bought Together