Ultrafiltration membrane filters, PES
These are general purpose membrane that provide excellent performance with most solutions when retentate recovery is of primary importance.
- No hydrophobic or hydrophilic interactions
- Low fouling characteristics, exceptional flux and broad pH range
- Catalog No:
- SART14629-25-D
- SART14650-76-D
- SART14679-47-D
- SART14659-25-D
- SART14679-76-D
- SART14609-47-D
- SART14639-25-D
- SART14659-47-D
- SART14650-47-D
- SART14639-47-D
- SART14668-47-D
- SART14629-47-D
- SART14629-76-D
- SART14609-76-D
- SART14639-76-D
- SART14629-63-D
- SART14650-25-D
- SART14609-63-D
- SART14659-63-D
- SART14639-63-D
- SART14668-63-D
- Thickness:
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
- 120 μm
Frequently Bought Together