Anti-CD163 Mouse Monoclonal Antibody
Catalog # PRSI51-296
Supplier: ProSci Inc.
Specifications
- Antigen name:CD163 molecule
- Antigen symbol:CD163
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:CD163 antigen|scavenger receptor cysteine-rich type 1 protein M130|hemoglobin scavenger receptor|macrophage-associated antigen
- Immunogen:Spleen cells from a case of hairy cell leukemia
- Tested applications:Fluorescence-Activated Cell Sorting
- Size:100 µg
- Pk:100 µG
Specifications
About this item
CD163 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.
Affinity Purified
PBS, pH 7.2, <=0.09% sodium azide
Store CD163 antibody at 4 °C. As with all antibodies avoid freeze/thaw cycles
Type:
Antigen: CD163
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human